Assay Details
| Gene Symbol: Mir1191b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106141.1 | 1 | 27 | 58 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nuclear factor I/C | Nfic |
1110019L22Rik, 1500041O16Rik, AA589446, AI746521, NF1-C |
Sense | Chr. 10 - 81396191 - 81431167 on NCBI Build GRCm38 | |
| Downstream |
CUGBP, Elav-like family member 5 |
Celf5 |
4930565A21Rik, Brunol5 |
42.9kb | Sense | Chr. 10 - 81459227 - 81482725 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir1191b
Genomic Map