Assay Details
| Gene Symbol: Tcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110341.1 | 2 - 3 | 326 | 86 | ||
| GenBank mRNA | CA558861.1 | - | 2 - 3 | 295 | 86 | |
| DV655843.1 | - | 2 - 3 | 319 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
T cell leukemia/lymphoma 1B, 4 |
Tcl1b4 | AA675342 | -11.2kb | Anti-sense | Chr. 12 - 105201957 - 105206993 on NCBI Build GRCm38 |
| Downstream | Tcl1 upstream neural differentiation associated RNA | Tunar |
2810011L19Rik, Tuna |
117.8kb | Anti-sense | Chr. 12 - 105336594 - 105383932 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Tcl1
Genomic Map