Assay Details
| Gene Symbol: Iqsec3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110358.1 | 13 - 14 | 3922 | 57 | ||
| GenBank mRNA | AK147351.1 | - | 13 - 14 | 3868 | 57 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 6 (neurotransmitter transporter, betaine/GABA), member 12 |
Slc6a12 |
BGT1, GAT2, Gabt2 |
-7.2kb | Anti-sense | Chr. 6 - 121342599 - 121365773 on NCBI Build GRCm38 |
| Downstream | alpha-2-macroglobulin | A2m | A2mp | 256.3kb | Anti-sense | Chr. 6 - 121636166 - 121679238 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Iqsec3
Genomic Map