Assay Details
| Gene Symbol: Lincenc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110430.1 | 1 - 2 | 77 | 79 | ||
| GenBank mRNA | BY736053.1 | - | 1 - 2 | 78 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 33447 |
Gm33447 | -100.5kb | Sense | Chr. 13 - 97376180 - 97382156 on NCBI Build GRCm38 | |
| Downstream | predicted gene 5086 | Gm5086 |
B230311A21, EG328314 |
77.0kb | Sense | Chr. 13 - 97560000 - 97583994 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Lincenc1
Genomic Map