Assay Details
| Gene Symbol: Lincpint | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110470.1 | 1 - 2 | 398 | 67 | ||
| GenBank mRNA | KC860258.1 | - | 1 - 2 | 270 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 29b-1 | Mir29b-1 |
Mirn29b, Mirn29b-1, mir-29b-1 |
-117.9kb | Sense | Chr. 6 - 31063023 - 31063093 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 2210408F21 gene | 2210408F21Rik | 1.9kb | Anti-sense | Chr. 6 - 31220351 - 31337395 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Lincpint
Genomic Map