Assay Details
| Gene Symbol: Gm38407 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110487.1 | 10 - 11 | 1892 | 96 | ||
| GenBank mRNA | AK029853.1 | - | 10 - 11 | 1894 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | retinol dehydrogenase 14 (all-trans and 9-cis) | Rdh14 |
3110030G19Rik, AW215439, PAN2 |
-117.3kb | Sense | Chr. 12 - 10390690 - 10395562 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 7420701I03 gene | 7420701I03Rik | 533.0kb | Sense | Chr. 12 - 11047813 - 11078741 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm38407
Genomic Map