Assay Details
| Gene Symbol: Gm12440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110516.1 | 3 - 4 | 857 | 79 | ||
| GenBank mRNA | AK076933.1 | - | 3 - 4 | 858 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | hydroxyacid oxidase 2 | Hao2 |
AI325478, Hao-2, Hao3, Haox3 |
-157.8kb | Anti-sense | Chr. 3 - 98874519 - 98895377 on NCBI Build GRCm38 |
| Downstream | tryptophanyl tRNA synthetase 2 (mitochondrial) | Wars2 |
5730427B17Rik, 9430020O07Rik, AI413375, TrpRS |
86.9kb | Sense | Chr. 3 - 99140618 - 99220203 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm12440
Genomic Map