Assay Details
| Gene Symbol: Slc39a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110981.1 | 3 - 4 | 405 | 131 | ||
| GenBank mRNA | BC020106.1 | - | 3 - 4 | 320 | 131 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
proteasome (prosome, macropain) 26S subunit, ATPase 3 |
Psmc3 | TBP-1 | -5.3kb | Anti-sense | Chr. 2 - 91054016 - 91059438 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 39874 |
Gm39874 | 4.7kb | Anti-sense | Chr. 2 - 91070420 - 91071623 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC020106.1 transcripts.
Set Membership: |
|

Slc39a13
Genomic Map