Assay Details
| Gene Symbol: Paupar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_117095.1 | 1 | 422 | 86 | ||
| GenBank mRNA | KJ434993.1 | - | 1 | 422 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | reticulocalbin 1 | Rcn1 | Rcn | -258.5kb | Sense | Chr. 2 - 105385948 - 105399319 on NCBI Build GRCm38 |
| Downstream | paired box 6 | Pax6 |
1500038E17Rik, AEY11, Dey, Gsfaey11, Pax-6, Sey |
6.2kb | Anti-sense | Chr. 2 - 105667518 - 105697364 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Paupar
Genomic Map