Assay Details
| Gene Symbol: Trim46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121683.1 | 1 - 2 | 311 | 113 | ||
| GenBank mRNA | BC065049.1 | - | 1 - 2 | 224 | 6418851 | 113 |
| CA315205.1 | - | 1 - 2 | 311 | 113 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
mucin 1, transmembrane |
Muc1 |
CD227, EMA, Muc-1 |
-10.8kb | Anti-sense | Chr. 3 - 89229056 - 89233381 on NCBI Build GRCm38 |
| Downstream | keratinocyte associated protein 2 | Krtcap2 |
0610010I12Rik, Kcp2 |
1.3kb | Anti-sense | Chr. 3 - 89246300 - 89249723 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC065049.1 transcripts.
Set Membership: |
|

Trim46
Genomic Map