Assay Details
| Gene Symbol: Trim46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121683.1 | 8 - 9 | 1579 | 67 | ||
| GenBank mRNA | BC065049.1 | - | 8 - 9 | 1492 | 6418851 | 67 |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
mucin 1, transmembrane |
Muc1 |
CD227, EMA, Muc-1 |
-4.2kb | Anti-sense | Chr. 3 - 89229056 - 89233381 on NCBI Build GRCm38 |
| Downstream | keratinocyte associated protein 2 | Krtcap2 |
0610010I12Rik, Kcp2 |
7.6kb | Anti-sense | Chr. 3 - 89246300 - 89249723 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Trim46
Genomic Map