Assay Details
| Gene Symbol: Ocel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_122030.1 | 1 - 2 | 718 | 103 | ||
| GenBank mRNA | BB766991.1 | - | 1 - 2 | 288 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | unconventional SNARE in the ER 1 homolog (S. cerevisiae) | Use1 |
2010315L10Rik, 5730403H22Rik, AV002165, D12, Ed2, Q-snare, p31 |
-1.6kb | Sense | Chr. 8 - 71366848 - 71369732 on NCBI Build GRCm38 |
| Downstream |
nuclear receptor subfamily 2, group F, member 6 |
Nr2f6 |
AV090102, COUP-TF3, EAR2, Erbal2 |
1.3kb | Anti-sense | Chr. 8 - 71374118 - 71381955 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Ocel1
Genomic Map