Assay Details
| Gene Symbol: A330069E16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015464.1 | 1 - 2 | 252 | 95 | ||
| GenBank mRNA | AK039596.1 | - | 1 - 2 | 202 | 95 | |
| BC042745.1 | - | 1 - 2 | 253 | 95 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | damage specific DNA binding protein 2 | Ddb2 | 2610043A19Rik | -14.0 | Anti-sense | Chr. 2 - 91211579 - 91237132 on NCBI Build GRCm38 |
| Downstream | protein kinase C and casein kinase substrate in neurons 3 | Pacsin3 |
4921507A02Rik, 6330413E15Rik, AW413130 |
17.7kb | Sense | Chr. 2 - 91256098 - 91264680 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

A330069E16Rik
Genomic Map