Assay Details
| Gene Symbol: Mir3473g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106202.1 | 1 | 84 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
transient receptor potential cation channel, subfamily M, member 7 |
Trpm7 |
2310022G15Rik, 4833414K03Rik, 5033407O22Rik, CHAK, CHAK1, LTrpC-7, Ltpr7, Ltrpc7, TRPPLIK |
-26.0kb | Sense | Chr. 2 - 126791558 - 126876261 on NCBI Build GRCm38 |
| Downstream |
adaptor-related protein complex AP-4, epsilon 1 |
Ap4e1 |
2310033A20Rik, 9930028M04Rik, AV087807 |
106.3kb | Anti-sense | Chr. 2 - 127008679 - 127069814 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir3473g
Genomic Map