Assay Details
| Gene Symbol: Psme3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011192.3 | NP_035322.1 | 9 - 10 | 791 | 100 | |
| GenBank mRNA | AK140597.1 | - | 9 - 10 | 812 | 100 | |
| AK150939.1 | - | 9 - 10 | 811 | 100 | ||
| AK156079.1 | - | 9 - 10 | 761 | 100 | ||
| AK171348.1 | - | 9 - 10 | 807 | 100 | ||
| BC005524.1 | - | 1 - 2 | 44 | 100 | ||
| BC015255.1 | - | 9 - 10 | 791 | 4221225 | 100 | |
| BC139818.1 | - | 9 - 10 | 739 | 40141268 | 100 | |
| CT010198.1 | - | 9 - 10 | 595 | 100 | ||
| D87911.1 | - | 9 - 10 | 714 | 100 | ||
More Information
Set Membership: |
|

Psme3
Genomic Map