Assay Details
| Gene Symbol: Egr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289925.1 | NP_001276854.1 | 2 - 3 | 206 | 77 | |
| GenBank mRNA | BC103568.1 | - | 2 - 3 | 204 | 40045218 | 77 |
| BC103570.1 | - | 2 - 3 | 205 | 40045221 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103568.1, BC103570.1, NM_001289925.1 transcripts.
Set Membership: |
|

Egr3
Genomic Map