Assay Details
| Gene Symbol: Pemt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290011.1 | NP_001276940.1 | 6 - 7 | 703 | 87 | |
| NM_001290012.1 | NP_001276941.1 | 6 - 7 | 699 | 87 | ||
| NM_001290013.1 | NP_001276942.1 | 6 - 7 | 637 | 87 | ||
| NM_001290014.1 | NP_001276943.1 | 4 - 5 | 379 | 87 | ||
| NM_008819.3 | NP_032845.2 | 6 - 7 | 737 | 87 | ||
| GenBank mRNA | AK133744.1 | - | 6 - 7 | 699 | 87 | |
| BC026796.1 | - | 6 - 7 | 571 | 4196324 | 87 | |
More Information
Set Membership: |
|

Pemt
Genomic Map