Assay Details
| Gene Symbol: Hax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282032.1 | NP_001268961.1 | 4 - 5 | 549 | 104 | |
| NM_011826.4 | NP_035956.1 | 5 - 6 | 801 | 104 | ||
| GenBank mRNA | AF023482.1 | - | 5 - 6 | 773 | 104 | |
| AF023483.1 | - | 4 - 5 | 936 | 104 | ||
| AF465243.1 | - | 4 - 5 | 413 | 104 | ||
| AK002256.1 | - | 5 - 6 | 752 | 104 | ||
| AK010633.1 | - | 5 - 6 | 785 | 104 | ||
| AK088746.1 | - | 5 - 6 | 783 | 104 | ||
| AK135138.1 | - | 4 - 5 | 511 | 104 | ||
| BC006688.1 | - | 5 - 6 | 730 | 3582652 | 104 | |
| BC098225.1 | - | 5 - 6 | 755 | 1494110 | 104 | |
| BC147481.1 | - | 5 - 6 | 732 | 9056226 | 104 | |
More Information
Set Membership: |
|

Hax1
Genomic Map