Assay Details
| Gene Symbol: Adipoq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009605.4 | NP_033735.3 | 2 - 3 | 336 | 77 | |
| GenBank mRNA | AK003138.1 | - | 2 - 3 | 254 | 77 | |
| AK134112.1 | - | 2 - 3 | 336 | 77 | ||
| AX767957.1 | - | 2 - 3 | 267 | 77 | ||
| AY749429.1 | - | 1 - 2 | 222 | 77 | ||
| AY754346.1 | - | 1 - 2 | 222 | 77 | ||
| BC028770.1 | - | 2 - 3 | 251 | 1329799 | 77 | |
| U37222.1 | - | 2 - 3 | 267 | 77 | ||
More Information
Set Membership: |
|

Adipoq
Genomic Map