Assay Details
| Gene Symbol: Gkap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019832.3 | NP_062806.1 | 2 - 3 | 376 | 80 | |
| GenBank mRNA | AB033130.1 | - | 1 - 2 | 235 | 80 | |
| AK016588.1 | - | 1 - 2 | 148 | 80 | ||
| AK140052.1 | - | 1 - 2 | 796 | 80 | ||
| BC052347.1 | - | 2 - 3 | 376 | 6774715 | 80 | |
More Information
Set Membership: |
|

Gkap1
Genomic Map