Assay Details
| Gene Symbol: Gkap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019832.3 | NP_062806.1 | 6 - 7 | 739 | 165 | |
| GenBank mRNA | AB033130.1 | - | 5 - 6 | 598 | 165 | |
| AK016588.1 | - | 5 - 6 | 511 | 165 | ||
| BC052347.1 | - | 6 - 7 | 739 | 6774715 | 165 | |
More Information
Set Membership: |
|

Gkap1
Genomic Map