Assay Details
| Gene Symbol: LOC105246506 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK141205.1 | - | 562 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ADNP homeobox 2 | Adnp2 |
8430420L05Rik, BC024969, Kiaa0863, Zfp508, mKIAA0863 |
-36.7kb | Anti-sense | Chr. 18 - 80126315 - 80152063 on NCBI Build GRCm38 |
| Downstream | ribosome binding factor A | Rbfa |
1110032A13Rik, AI595940 |
2.1kb | Anti-sense | Chr. 18 - 80192264 - 80200619 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

LOC105246506
Genomic Map