Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 29683 |
Gm29683 | Sense | Chr. 7 - 70267693 - 70339083 on NCBI Build GRCm38 | ||
| Downstream |
nuclear receptor subfamily 2, group F, member 2 |
Nr2f2 |
2700033K02Rik, 9430015G03Rik, ARP-1, Aporp1, COUP-TF2, COUP-TFII, COUPTFB, EAR3, SVP40, Tcfcoup2 |
14.1kb | Sense | Chr. 7 - 70351946 - 70366746 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map