Assay Details
| Gene Symbol: Cryz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009968.3 | NP_034098.1 | 9 | 1984 | 99 | |
| GenBank mRNA | AK098082.1 | - | 9 | 1468 | 99 | |
| AK133820.1 | - | 10 | 2084 | 99 | ||
| AK134281.1 | - | 9 | 1469 | 99 | ||
| BC003800.1 | - | 9 | 1475 | 3492859 | 99 | |
| BC043076.1 | - | 9 | 1445 | 6404996 | 99 | |
| D78646.1 | - | 6 | 1011 | 99 | ||
| S70056.1 | - | 9 | 1496 | 99 | ||
More Information
Set Membership: |
|

Cryz
Genomic Map