Assay Details
| Gene Symbol: Zfp652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201609.2 | NP_963903.2 | 1 | 485 | 92 | |
| GenBank mRNA | AK220361.1 | - | 1 | 330 | 92 | |
| BC055752.1 | - | 1 | 485 | 6414140 | 92 | |
More Information
Set Membership: |
|

Zfp652
Genomic Map