Assay Details
| Gene Symbol: Adamts14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081127.1 | NP_001074596.1 | 22 | 4667 | 92 | |
| GenBank mRNA | AK137573.1 | - | 7 | 2119 | 92 | |
| BC157953.1 | - | 22 | 4488 | 9007283 | 92 | |
| BC158014.1 | - | 22 | 4500 | 9088151 | 92 | |
| BC172106.1 | - | 22 | 4481 | 92 | ||
More Information
Set Membership: |
|

Adamts14
Genomic Map