Assay Details
| Gene Symbol: Luc7l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK084533.1 | - | 1 | 312 | 103 | |
| AK163764.1 | - | 1 | 2837 | 103 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Luc7l2| Gene Symbol: Luc7l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK084533.1 | - | 1 | 312 | 103 | |
| AK163764.1 | - | 1 | 2837 | 103 | ||
Set Membership: |
|