Assay Details
| Gene Symbol: Ap3m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018829.4 | NP_061299.3 | 9 | 2106 | 78 | |
| GenBank mRNA | AK004740.1 | - | 9 | 1852 | 78 | |
| BC010667.1 | - | 3 | 989 | 78 | ||
| BC024595.1 | - | 9 | 1820 | 5026206 | 78 | |
| BC090983.1 | - | 9 | 1854 | 30282230 | 78 | |
More Information
Set Membership: |
|

Ap3m1
Genomic Map