Assay Details
| Gene Symbol: Dedd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK043908.1 | - | 3853 | 97 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dedd2| Gene Symbol: Dedd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK043908.1 | - | 3853 | 97 | ||