Assay Details
| Gene Symbol: A730052K04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK043070.1 | - | 1 | 215 | 82 | |
| BB640342.1 | - | 1 | 215 | 82 | ||
| Gene Symbol: Ushbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK034939.1 | - | 6 | 82 | ||
More Information
Set Membership: |
|

A730052K04Rik
Genomic Map