Assay Details
| Gene Symbol: Rab8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023126.2 | NP_075615.2 | 8 | 1522 | 84 | |
| GenBank mRNA | AK003319.1 | - | 1 | 691 | 84 | |
| AK011128.1 | - | 1 | 203 | 84 | ||
| AK076048.1 | - | 8 | 1401 | 84 | ||
| AK079306.1 | - | 1389 | 84 | |||
| AK080740.1 | - | 8 | 1374 | 84 | ||
| AK147178.1 | - | 9 | 840 | 84 | ||
More Information
Set Membership: |
|

Rab8a
Genomic Map