Assay Details
| Gene Symbol: Nfatc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010901.2 | NP_035031.2 | 10 | 4075 | 92 | |
| GenBank mRNA | AK088536.1 | - | 2 | 927 | 92 | |
| AK142470.1 | - | 1 | 269 | 92 | ||
| AK161185.1 | - | 2 | 1125 | 92 | ||
More Information
Set Membership: |
|

Nfatc3
Genomic Map