Assay Details
| Gene Symbol: Zfp276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK083110.1 | - | 2164 | 90 | ||
| BC078415.1 | - | 10 | 1486 | 90 | ||
| BC145340.1 | - | 9 | 1094 | 90 | ||
| Gene Symbol: Fanca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK054319.1 | - | 90 | |||
More Information
Set Membership: |
|

Zfp276
Genomic Map