Assay Details
| Gene Symbol: Zfp276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK083110.1 | - | 1867 | 100 | ||
| Gene Symbol: Fanca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK054319.1 | - | 100 | |||

Zfp276
Genomic Map