Assay Details
| Gene Symbol: 4833445I07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076537.1 | - | 1 | 1214 | 82 | |
| AV253134.2 | - | 1 | 224 | 82 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

4833445I07Rik| Gene Symbol: 4833445I07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076537.1 | - | 1 | 1214 | 82 | |
| AV253134.2 | - | 1 | 224 | 82 | ||
Set Membership: |
|