Assay Details
| Gene Symbol: Eif4e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007917.3 | NP_031943.3 | 7 | 653 | 109 | |
| GenBank mRNA | AK082867.1 | - | 6 | 109 | ||
| AK146757.1 | - | 6 | 533 | 109 | ||
| AK167097.1 | - | 6 | 542 | 109 | ||
| BC010759.1 | - | 6 | 527 | 4216609 | 109 | |
| BC085087.1 | - | 6 | 525 | 6438419 | 109 | |
| M61731.1 | - | 6 | 539 | 109 | ||
More Information
Set Membership: |
|

Eif4e
Genomic Map