Assay Details
| Gene Symbol: Pibf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029320.3 | NP_083596.2 | 4 | 1198 | 92 | |
| GenBank mRNA | AK015781.1 | - | 4 | 665 | 92 | |
| AK017107.1 | - | 4 | 1199 | 92 | ||
| AK030385.1 | - | 4 | 679 | 92 | ||
| AK045133.1 | - | 4 | 693 | 92 | ||
| BC113804.1 | - | 4 | 675 | 92 | ||
| BC129811.1 | - | 4 | 675 | 40062382 | 92 | |
| BC129812.1 | - | 4 | 675 | 40062383 | 92 | |
More Information
Set Membership: |
|

Pibf1
Genomic Map