Assay Details
| Gene Symbol: Hs3st3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018805.2 | NP_061275.2 | 2 | 5181 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hs3st3b1| Gene Symbol: Hs3st3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018805.2 | NP_061275.2 | 2 | 5181 | 90 | |
Set Membership: |
|