Assay Details
| Gene Symbol: Tlcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK046577.1 | - | 2 | 559 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK046577.1 transcripts.
Set Membership: |
|

Tlcd1
Genomic Map