Assay Details
| Gene Symbol: Tlcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK161721.1 | - | 1842 | 65 | ||
| AK165432.1 | - | 2054 | 65 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tlcd1| Gene Symbol: Tlcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK161721.1 | - | 1842 | 65 | ||
| AK165432.1 | - | 2054 | 65 | |||