Assay Details
| Gene Symbol: Rhob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007483.2 | NP_031509.1 | 1 | 1060 | 83 | |
| GenBank mRNA | AF481943.1 | - | 1 | 707 | 83 | |
| AK013784.1 | - | 1 | 232 | 83 | ||
| AK148834.1 | - | 1 | 138 | 83 | ||
| AK164518.1 | - | 1 | 1085 | 83 | ||
| AK167917.1 | - | 1 | 1085 | 83 | ||
| AK215498.1 | - | 1 | 50 | 83 | ||
| BC018275.1 | - | 1 | 1060 | 4168565 | 83 | |
| BC023334.1 | - | 1 | 816 | 83 | ||
| BC110992.1 | - | 1 | 1071 | 5376565 | 83 | |
More Information
Set Membership: |
|

Rhob
Genomic Map