Assay Details
| Gene Symbol: Gm15564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK144265.1 | - | 182 | 88 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15564| Gene Symbol: Gm15564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK144265.1 | - | 182 | 88 | ||