Assay Details
| Gene Symbol: Six3os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY589792.1 | - | 60 | 71 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Six3os1| Gene Symbol: Six3os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY589792.1 | - | 60 | 71 | ||