Assay Details
| Gene Symbol: Sh3pxd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164717.1 | NP_001158189.1 | 14 | 6635 | 70 | |
| NM_008018.4 | NP_032044.2 | 15 | 6719 | 70 | ||
More Information
Set Membership: |
|

Sh3pxd2a
Genomic Map