Assay Details
| Gene Symbol: Rab11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008997.3 | NP_033023.1 | 4 - 5 | 740 | 66 | |
| GenBank mRNA | AB232606.1 | - | 4 - 5 | 516 | 66 | |
| AK134125.1 | - | 4 - 5 | 485 | 66 | ||
| AK146452.1 | - | 4 - 5 | 601 | 66 | ||
| AK146460.1 | - | 4 - 5 | 552 | 66 | ||
| AK146623.1 | - | 4 - 5 | 552 | 66 | ||
| AK146978.1 | - | 4 - 5 | 532 | 66 | ||
| AK147077.1 | - | 4 - 5 | 571 | 66 | ||
| AK148022.1 | - | 4 - 5 | 613 | 66 | ||
| AK148106.1 | - | 4 - 5 | 539 | 66 | ||
| AK188721.1 | - | 2 - 3 | 189 | 66 | ||
| AK203890.1 | - | 2 - 3 | 214 | 66 | ||
| AK205000.1 | - | 2 - 3 | 214 | 66 | ||
| BC054753.1 | - | 4 - 5 | 556 | 6411651 | 66 | |
| BC085270.1 | - | 4 - 5 | 549 | 6478784 | 66 | |
| BC157916.1 | - | 4 - 5 | 569 | 9007344 | 66 | |
| KJ650201.1 | - | 4 - 5 | 516 | 66 | ||
| L26528.1 | - | 4 - 5 | 551 | 66 | ||
More Information
Set Membership: |
|

Rab11b
Genomic Map