Assay Details
| Gene Symbol: Ap2m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK141862.1 | - | 3 | 411 | 74 | |
| AK143586.1 | - | 3 | 426 | 74 | ||
| AK146517.1 | - | 3 | 480 | 74 | ||
| AK150358.1 | - | 3 | 447 | 74 | ||
| AK159534.1 | - | 3 | 458 | 74 | ||
| AK168414.1 | - | 3 | 457 | 74 | ||
| AK172889.1 | - | 3 | 411 | 74 | ||
| BC056352.1 | - | 3 | 450 | 6807694 | 74 | |
| BC089342.1 | - | 3 | 443 | 30538471 | 74 | |
| BC094510.1 | - | 3 | 440 | 6313261 | 74 | |
| U27106.1 | - | 3 | 456 | 74 | ||
More Information
Set Membership: |
|

Ap2m1
Genomic Map