Assay Details
| Gene Symbol: Usf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF479773.1 | - | 7 | 701 | 103 | |
| AK142437.1 | - | 9 | 954 | 103 | ||
| BC049784.1 | - | 9 | 897 | 6506829 | 103 | |
More Information
Set Membership: |
|

Usf1
Genomic Map