Assay Details
| Gene Symbol: Mafg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047224.1 | - | 1766 | 123 | ||
| AK083165.1 | - | 2343 | 123 | |||
| AK194292.1 | - | 126 | ||||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Mafg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047224.1 | - | 1766 | 123 | ||
| AK083165.1 | - | 2343 | 123 | |||
| AK194292.1 | - | 126 | ||||
Set Membership: |
|