Assay Details
| Gene Symbol: Foxo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019739.3 | NP_062713.2 | 1 | 371 | 116 | |
| GenBank mRNA | AF114258.1 | - | 1 | 289 | 116 | |
| AJ252157.1 | - | 1 | 334 | 116 | ||
| AK137629.1 | - | 1 | 319 | 116 | ||
| AK154041.1 | - | 1 | 372 | 116 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF114258.1, AJ252157.1, AK137629.1, AK154041.1, NM_019739.3 transcripts.
Set Membership: |
|

Foxo1
Genomic Map