Assay Details
| Gene Symbol: Tiparp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178892.5 | NP_849223.2 | 6 | 3271 | 96 | |
| GenBank mRNA | AK028529.1 | - | 6 | 3273 | 96 | |
| BC068173.1 | - | 6 | 3245 | 30616032 | 96 | |
More Information
Set Membership: |
|

Tiparp
Genomic Map